Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rap1b
Entrez Gene ID
171337
Gene Name
RAP1B, member of RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 60850406 - 60878225 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008765377.1 XP_008763599.1 6 - 7 1020 154
GenBank mRNA BC081731.1 - 5 - 6 605 154
FQ229455.1 - 5 - 6 634 154
U07795.1 - 5 - 6 540 154

View Full Product Details