Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Metap2
- Entrez Gene ID
-
64370
- Gene Name
- methionyl aminopeptidase 2
- Gene Aliases
-
Amp2, Mnpep, p67
- Chromosome Location
-
Chr.7: 34762241 - 34789456 on Build Rnor_6.0
- Gene Symbol
- RGD1560341
- Entrez Gene ID
-
498869
- Gene Name
- similar to Methionine aminopeptidase 2 (MetAP 2)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 51218243 - 51220149 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_574154.6 |
XP_574154.2 |
1 - 1 |
443 |
68 |
View Full Product Details