The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007750.1 | NP_001007751.1 | 7 - 8 | 1161 | 148 |
XM_006241228.2 | XP_006241290.1 | 7 - 8 | 1156 | 148 | |
XM_006241230.2 | XP_006241292.1 | 7 - 8 | 1156 | 148 | |
XM_006241231.2 | XP_006241293.1 | 7 - 8 | 1156 | 148 | |
GenBank mRNA | BC082074.1 | - | 7 - 8 | 1161 | 148 |