Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmtc2
Entrez Gene ID
299762
Gene Name
transmembrane and tetratricopeptide repeat containing 2
Gene Aliases
RGD1309848
Chromosome Location
Chr.7: 47181439 - 47586137 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171177.2 NP_001164648.2 3 - 4 1483 116

View Full Product Details