The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108081.1 | NP_001101551.1 | 10 - 11 | 1119 | 100 |
XM_006241243.1 | XP_006241305.1 | 10 - 11 | 1128 | 100 | |
GenBank mRNA | BC166860.1 | - | 10 - 11 | 1095 | 100 |
FQ220136.1 | - | 7 - 8 | 763 | 100 |