The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001055430.5 | XP_001055430.4 | 5 - 6 | 945 | 74 |
XM_006241414.2 | XP_006241476.1 | 6 - 7 | 539 | 74 | |
XM_008765427.1 | XP_008763649.1 | 5 - 6 | 965 | 74 | |
XM_008765428.1 | XP_008763650.1 | 6 - 7 | 589 | 74 |