Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acsm5
Entrez Gene ID
361637
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
Aa2-174, Cc1-38, RGD1309578
Chromosome Location
Chr.1: 189241364 - 189270422 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014162.4 NP_001014184.3 5 - 6 1008 88
XM_006230123.1 XP_006230185.1 6 - 7 951 88
XM_006230126.1 XP_006230188.1 6 - 7 951 88
GenBank mRNA AY325244.1 - - 1244 88
BC078915.1 - 5 - 6 1007 88

View Full Product Details