Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gsg1l
Entrez Gene ID
499263
Gene Name
GSG1-like
Gene Aliases
RGD1562278
Chromosome Location
Chr.1: 197271704 - 197482622 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759923.1 XP_008758145.1 5 - 6 898 77

View Full Product Details