Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctf1
Entrez Gene ID
29201
Gene Name
cardiotrophin 1
Gene Aliases
-
Chromosome Location
Chr.1: 199162320 - 199168299 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017129.1 NP_058825.1 1 - 2 61 70
GenBank mRNA BC072690.1 - 1 - 2 50 70
D78591.1 - 1 - 2 61 70

View Full Product Details