Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spns1
Entrez Gene ID
361648
Gene Name
spinster homolog 1 (Drosophila)
Gene Aliases
RGD1305613
Chromosome Location
Chr.1: 197771196 - 197778475 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039208.2 NP_001034297.2 9 - 10 1570 67

View Full Product Details