Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Parva
Entrez Gene ID
57341
Gene Name
parvin, alpha
Gene Aliases
Actp
Chromosome Location
Chr.1: 177249302 - 177405143 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020656.2 NP_065707.2 4 - 5 398 83
GenBank mRNA AF264765.2 - 4 - 5 398 83
FQ228521.1 - - 458 83

View Full Product Details