The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277302.1 | NP_001264231.1 | 1 - 2 | 402 | 60 |
GenBank mRNA | FQ212642.1 | - | 1 - 2 | 391 | 60 |
FQ213714.1 | - | 1 - 2 | 402 | 60 | |
FQ229379.1 | - | 1 - 2 | 76 | 60 | |
FQ230130.1 | - | 1 - 2 | 390 | 60 |