Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyve1
Entrez Gene ID
293186
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Gene Aliases
Xlkd1
Chromosome Location
Chr.1: 175663266 - 175676699 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106286.1 NP_001099756.1 1 - 2 447 73

View Full Product Details