Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iqck
Entrez Gene ID
100909537
Gene Name
IQ motif containing K
Gene Aliases
-
Chromosome Location
Chr.1: 188571934 - 188690184 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230149.2 XP_006230211.1 8 - 9 823 75

View Full Product Details