Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nucb2
Entrez Gene ID
59295
Gene Name
nucleobindin 2
Gene Aliases
Nefa, p54
Chromosome Location
Chr.1: 185106773 - 185143276 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021663.2 NP_067695.1 8 - 9 964 112
XM_006230095.2 XP_006230157.1 7 - 8 872 112
GenBank mRNA AF238223.1 - 6 - 7 751 112
AF250142.1 - 7 - 8 851 112
BC061778.1 - 8 - 9 964 112

View Full Product Details