The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024750.1 | NP_001019921.1 | 11 - 12 | 1280 | 117 |
GenBank mRNA | BC098010.1 | - | 11 - 12 | 1280 | 117 |
FQ211238.1 | - | - | 1281 | 117 |