Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rabep2
Entrez Gene ID
80754
Gene Name
rabaptin, RAB GTPase binding effector protein 2
Gene Aliases
Fra, MP13
Chromosome Location
Chr.1: 197839583 - 197855953 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030585.1 NP_085074.1 12 - 13 1625 72
GenBank mRNA BC089795.1 - 12 - 13 1611 72
U34932.1 - 12 - 13 1625 72

View Full Product Details