Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btbd10
Entrez Gene ID
308890
Gene Name
BTB (POZ) domain containing 10
Gene Aliases
RGD1306301
Chromosome Location
Chr.1: 178138562 - 178196551 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014022.1 NP_001014044.1 5 - 6 759 96
XM_006230033.2 XP_006230095.1 4 - 5 2214 96
XM_006230034.2 XP_006230096.1 7 - 8 1239 96
XM_006230035.2 XP_006230097.1 6 - 7 1210 96
XM_006230036.2 XP_006230098.1 6 - 7 1174 96
XM_006230038.2 XP_006230100.1 5 - 6 757 96
XM_006230040.2 XP_006230102.1 4 - 5 1033 96
XM_006230041.2 XP_006230103.1 6 - 7 1081 96
XM_006230043.2 XP_006230105.1 4 - 5 3340 96
XM_008759724.1 XP_008757946.1 5 - 6 2299 96
XM_008759725.1 XP_008757947.1 5 - 6 1129 96
XM_008759726.1 XP_008757948.1 4 - 5 2214 96
GenBank mRNA BC090330.1 - 5 - 6 759 96

View Full Product Details