Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grm5
Entrez Gene ID
24418
Gene Name
glutamate receptor, metabotropic 5
Gene Aliases
mGluR5, mGlur5
Chromosome Location
Chr.1: 151207859 - 151784991 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229659.2 XP_006229721.1 9 - 10 3314 131
GenBank mRNA EU559292.1 - 8 - 9 2720 131

View Full Product Details