The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106296.2 | NP_001099766.2 | 4 - 5 | 704 | 66 |
XM_006230237.2 | XP_006230299.1 | 5 - 6 | 780 | 66 | |
XM_006230239.2 | XP_006230301.1 | 4 - 5 | 525 | 66 | |
XM_006230240.2 | XP_006230302.1 | 4 - 5 | 1300 | 66 | |
GenBank mRNA | BC161920.1 | - | 4 - 5 | 704 | 66 |