The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053869.1 | NP_446321.1 | 2 - 3 | 603 | 76 |
GenBank mRNA | U25967.1 | - | 2 - 3 | 603 | 76 |