Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prss53
Entrez Gene ID
499270
Gene Name
protease, serine, 53
Gene Aliases
Pol3s, RGD1311277, RGD1566127
Chromosome Location
Chr.1: 199331108 - 199337123 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109156.1 NP_001102626.1 1 - 2 214 149
XM_006230322.2 XP_006230384.1 2 - 3 738 149
XM_006230324.2 XP_006230386.1 1 - 2 885 149
XM_008759914.1 XP_008758136.1 1 - 2 886 149
GenBank mRNA BC161977.1 - 1 - 2 177 149

View Full Product Details