The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109156.1 | NP_001102626.1 | 6 - 7 | 1044 | 119 |
XM_006230322.2 | XP_006230384.1 | 6 - 7 | 1685 | 119 | |
XM_006230324.2 | XP_006230386.1 | 6 - 7 | 1715 | 119 | |
XM_008759914.1 | XP_008758136.1 | 5 - 6 | 1833 | 119 | |
GenBank mRNA | BC161977.1 | - | 5 - 6 | 1154 | 119 |