The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182674.3 | NP_872615.3 | 4 - 5 | 768 | 102 |
GenBank mRNA | AY257675.4 | - | 4 - 5 | 751 | 102 |
BC079030.1 | - | 4 - 5 | 757 | 102 |