The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053537.2 | NP_445989.2 | 9 - 10 | 1604 | 66 |
GenBank mRNA | AY816233.1 | - | 9 - 10 | 1604 | 66 |
L27651.1 | - | 9 - 10 | 1664 | 66 |