Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tfeb
Entrez Gene ID
316214
Gene Name
transcription factor EB
Gene Aliases
Tcfeb
Chromosome Location
Chr.9: 15208141 - 15263966 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025707.1 NP_001020878.1 5 - 6 976 76
XM_006244440.2 XP_006244502.1 6 - 7 1905 76
XM_006244442.1 XP_006244504.1 6 - 7 895 76
XM_006244446.2 XP_006244508.1 6 - 7 2166 76
GenBank mRNA BC099102.1 - 5 - 6 976 76

View Full Product Details