Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vegfa
Entrez Gene ID
83785
Gene Name
vascular endothelial growth factor A
Gene Aliases
VEGF-A, VEGF164, VPF, Vegf
Chromosome Location
Chr.9: 17340341 - 17355681 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110333.2 NP_001103803.1 3 - 4 1328 95
NM_001110334.2 NP_001103804.1 3 - 4 1328 95
NM_001287107.1 NP_001274036.1 3 - 4 1328 95
NM_001287108.1 NP_001274037.1 3 - 4 1328 95
NM_001287110.1 NP_001274039.1 3 - 4 1328 95
NM_001287111.1 NP_001274040.1 3 - 4 1328 95
NM_001287112.1 NP_001274041.1 3 - 4 1328 95
NM_001287113.1 NP_001274042.1 2 - 3 1279 95
NM_001287114.1 NP_001274043.1 2 - 3 1279 95
NM_031836.3 NP_114024.2 3 - 4 1328 95
NR_105011.1 3 - 4 1328 95
GenBank mRNA AF215725.1 - 3 - 4 314 95
AF215726.1 - 3 - 4 95
AF222779.1 - 3 - 4 314 95
AY033503.1 - 2 - 3 276 95
AY033504.1 - 2 - 3 276 95
AY033506.1 - 3 - 4 325 95
AY033507.1 - 3 - 4 325 95
AY033508.1 - 3 - 4 325 95
AY702972.1 - 3 - 4 314 95
BC168708.1 - 3 - 4 1204 95
GQ423618.1 - 2 - 3 236 95
L20913.1 - 1 - 2 198 95
M32167.1 - 3 - 4 318 95

View Full Product Details