Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crisp4
Entrez Gene ID
654517
Gene Name
cysteine-rich secretory protein 4
Gene Aliases
-
Chromosome Location
Chr.9: 24356577 - 24383679 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039393.2 NP_001034482.2 3 - 4 374 70
XM_008766935.1 XP_008765157.1 3 - 4 374 70
GenBank mRNA DQ200907.1 - 3 - 4 374 70

View Full Product Details