Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfap2b
Entrez Gene ID
301285
Gene Name
transcription factor AP-2 beta
Gene Aliases
Tcfap2b
Chromosome Location
Chr.9: 25410669 - 25440426 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106896.2 NP_001100366.2 4 - 5 1072 112
XM_006244649.1 XP_006244711.1 4 - 5 1072 112
XM_008766927.1 XP_008765149.1 5 - 6 1051 112

View Full Product Details