Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnq5
Entrez Gene ID
259273
Gene Name
potassium channel, voltage-gated KQT-like subfamily Q, member 5
Gene Aliases
Kcnq5l
Chromosome Location
Chr.9: 27565869 - 28141114 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134643.2 NP_001128115.2 8 - 9 1393 117

View Full Product Details