Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc3
Entrez Gene ID
363192
Gene Name
kelch domain containing 3
Gene Aliases
-
Chromosome Location
Chr.9: 16612431 - 16618761 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012203.1 NP_001012203.1 5 - 6 742 77
XM_006244542.2 XP_006244604.1 4 - 5 592 77
GenBank mRNA BC079035.1 - 5 - 6 742 77

View Full Product Details