The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017382.1 | NP_001017382.1 | 4 - 5 | 487 | 82 |
XM_006244436.2 | XP_006244498.1 | 5 - 6 | 1004 | 82 | |
XM_006244437.2 | XP_006244499.1 | 5 - 6 | 858 | 82 | |
GenBank mRNA | AY972083.1 | - | 4 - 5 | 487 | 82 |