Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mocs1
Entrez Gene ID
301221
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
-
Chromosome Location
Chr.9: 13493705 - 13520113 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106881.1 NP_001100351.1 10 - 11 1225 133
XM_006244415.2 XP_006244477.1 11 - 12 1295 133
XM_006244416.2 XP_006244478.1 10 - 11 1236 133
XM_008766840.1 XP_008765062.1 7 - 8 643 133
XM_008766842.1 XP_008765064.1 5 - 6 537 133

View Full Product Details