Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcan2
Entrez Gene ID
140666
Gene Name
regulator of calcineurin 2
Gene Aliases
Dcip2, Dscr1l1, Zaki-4
Chromosome Location
Chr.9: 19518568 - 19749145 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175578.3 NP_783168.1 3 - 4 629 74
XM_006244593.2 XP_006244655.1 2 - 3 553 74
GenBank mRNA AF459022.1 - 2 - 3 351 74
AF459023.1 - 3 - 4 471 74
BC086974.1 - 3 - 4 629 74

View Full Product Details