The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108197.1 | NP_001101667.1 | 6 - 7 | 654 | 75 |
XM_008766860.1 | XP_008765082.1 | 6 - 7 | 655 | 75 | |
XM_008766861.1 | XP_008765083.1 | 6 - 7 | 655 | 75 | |
XM_008766862.1 | XP_008765084.1 | 6 - 7 | 655 | 75 | |
GenBank mRNA | FQ211123.1 | - | 6 - 7 | 651 | 75 |