Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptcra
Entrez Gene ID
116462
Gene Name
pre T-cell antigen receptor alpha
Gene Aliases
-
Chromosome Location
Chr.9: 16528972 - 16539378 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750652.3 XP_003750700.1 1 - 2 667 87

View Full Product Details