Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdca7
Entrez Gene ID
311742
Gene Name
cell division cycle associated 7
Gene Aliases
-
Chromosome Location
Chr.3: 59153281 - 59163916 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025693.1 NP_001020864.1 5 - 6 766 57
GenBank mRNA BC098690.1 - 5 - 6 766 57

View Full Product Details