The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047857.3 | NP_001041322.2 | 5 - 6 | 904 | 102 |
XM_006234405.2 | XP_006234467.1 | 5 - 6 | 831 | 102 | |
GenBank mRNA | DQ480743.1 | - | 5 - 6 | 907 | 102 |