Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spint1
Entrez Gene ID
311331
Gene Name
serine peptidase inhibitor, Kunitz type 1
Gene Aliases
HAI-1
Chromosome Location
Chr.3: 111049082 - 111061991 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004265.2 NP_001004265.2 7 - 8 1327 75
XM_006234763.1 XP_006234825.1 8 - 9 1339 75
XM_006234764.2 XP_006234826.1 8 - 9 1265 75
XM_008762100.1 XP_008760322.1 8 - 9 1339 75
GenBank mRNA AB154834.1 - 7 - 8 1100 75
BC097253.1 - 7 - 8 1327 75

View Full Product Details