The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107778.1 | NP_001101248.1 | 9 - 10 | 1080 | 59 |
XM_006235027.2 | XP_006235089.1 | 10 - 11 | 1235 | 59 | |
XM_006235028.2 | XP_006235090.1 | 9 - 10 | 1115 | 59 | |
XM_008762178.1 | XP_008760400.1 | 9 - 10 | 2156 | 59 |