Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nop10
Entrez Gene ID
691534
Gene Name
NOP10 ribonucleoprotein
Gene Aliases
Nola3
Chromosome Location
Chr.3: 103747654 - 103748739 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126100.1 NP_001119572.1 1 - 2 149 66

View Full Product Details