The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106509.1 | NP_001099979.1 | 2 - 3 | 290 | 87 |
XM_006234969.2 | XP_006235031.1 | 2 - 3 | 335 | 87 | |
GenBank mRNA | BC158612.1 | - | 2 - 3 | 192 | 87 |