Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scrn3
Entrez Gene ID
311731
Gene Name
secernin 3
Gene Aliases
-
Chromosome Location
Chr.3: 60023974 - 60048746 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013162.1 NP_001013180.1 3 - 4 390 111
XM_006234358.2 XP_006234420.1 3 - 4 427 111
XM_006234359.2 XP_006234421.1 3 - 4 429 111
XM_006234360.2 XP_006234422.1 3 - 4 429 111
GenBank mRNA BC088234.1 - 3 - 4 390 111

View Full Product Details