Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scrn3
Entrez Gene ID
311731
Gene Name
secernin 3
Gene Aliases
-
Chromosome Location
Chr.3: 60023974 - 60048746 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234358.2 XP_006234420.1 7 - 8 1170 74

View Full Product Details