Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcna4
Entrez Gene ID
25469
Gene Name
potassium channel, voltage gated shaker related subfamily A, member 4
Gene Aliases
KCHAN, Kv1.4, Kv4, RHK1, RK3
Chromosome Location
Chr.3: 98293298 - 98300763 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234673.2 XP_006234735.1 1 - 2 504 129

View Full Product Details