The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014047.1 | NP_001014069.1 | 2 - 3 | 359 | 134 |
XM_008762166.1 | XP_008760388.1 | 7 - 8 | 837 | 134 | |
XM_008762167.1 | XP_008760389.1 | 5 - 6 | 367 | 134 | |
XM_008762168.1 | XP_008760390.1 | 3 - 4 | 268 | 134 | |
XM_008762169.1 | XP_008760391.1 | 6 - 7 | 573 | 134 | |
GenBank mRNA | BC089959.1 | - | 2 - 3 | 359 | 134 |