Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casc5
Entrez Gene ID
311327
Gene Name
cancer susceptibility candidate 5
Gene Aliases
Rad51
Chromosome Location
Chr.3: 110847318 - 110909807 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170594.1 NP_001164065.1 13 - 14 5417 129
XM_008762095.1 XP_008760317.1 16 - 17 5603 129
XM_008762096.1 XP_008760318.1 16 - 17 5833 129
XM_008762097.1 XP_008760319.1 16 - 17 5646 129
XM_008762098.1 XP_008760320.1 14 - 15 5516 129
XM_008762099.1 XP_008760321.1 16 - 17 5590 129

View Full Product Details