The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111294.1 | NP_001104764.1 | 9 - 10 | 1163 | 69 |
NM_001111295.1 | NP_001104765.1 | 9 - 10 | 1097 | 69 | |
NM_198132.3 | NP_937765.1 | 9 - 10 | 1163 | 69 | |
XM_008761911.1 | XP_008760133.1 | 9 - 10 | 1112 | 69 | |
XM_008761912.1 | XP_008760134.1 | 9 - 10 | 1112 | 69 | |
GenBank mRNA | AY363226.1 | - | 9 - 10 | 1109 | 69 |
AY363227.1 | - | 9 - 10 | 1043 | 69 | |
BC081878.1 | - | 9 - 10 | 1103 | 69 | |
CF110891.1 | - | 3 - 4 | 329 | 69 | |
FQ214617.1 | - | 9 - 10 | 1059 | 69 | |
Y16641.1 | - | 5 - 6 | 525 | 69 |