Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fsip1
Entrez Gene ID
296074
Gene Name
fibrous sheath interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 109967302 - 110021184 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013078.1 NP_001013096.1 6 - 7 839 137
XM_006234734.2 XP_006234796.1 6 - 7 877 137
XM_006234735.2 XP_006234797.1 6 - 7 874 137
XM_008762090.1 XP_008760312.1 7 - 8 820 137
GenBank mRNA BC082080.1 - 6 - 7 839 137

View Full Product Details