Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acp2
Entrez Gene ID
24162
Gene Name
acid phosphatase 2, lysosomal
Gene Aliases
LAP
Chromosome Location
Chr.3: 80020347 - 80030365 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016988.2 NP_058684.2 8 - 9 898 80
XM_006234500.1 XP_006234562.1 9 - 10 1820 80
GenBank mRNA BC081823.1 - 8 - 9 869 80
M27893.1 - 8 - 9 867 80

View Full Product Details